Anti-TRIB1 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Lieferant: United States Biological
USBIT8335-40B-APEA 852 EUR
USBIT8335-40B-AP
Anti-TRIB1 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500
Do not freeze!
Type: Primary
Antigen: TRIB1 (tribbles pseudokinase 1)
Clonality: polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human