Anti-RNF216 Rabbit Polyclonal Antibody (FITC (Fluorescein))

Lieferant: United States Biological
USBIT8319-04A-FITCEA 852 EUR
USBIT8319-04A-FITC
Anti-RNF216 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-1:500; Immunochemistry: 1:50-1:100
Do not freeze!

Type: Primary
Antigen: RNF216 (ring finger protein 216)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR