Anti-RNF216 Rabbit Polyclonal Antibody (Biotin)

Lieferant: United States Biological
USBIT8319-04A-BIOTEA 852 EUR
USBIT8319-04A-BIOT
Anti-RNF216 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: RNF216 (ring finger protein 216)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR