Anti-PAPD7 Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBIT8302-50EA 507 EUR
USBIT8302-50
Anti-PAPD7 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PAPD7 (PAP associated domain containing 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR