Anti-Treponema pallidum Rabbit Polyclonal Antibody (FITC (Fluorescein))

Lieferant: United States Biological
USBIT8300-11CEA 460 EUR
USBIT8300-11C
Anti-Treponema pallidum Rabbit Polyclonal Antibody (FITC (Fluorescein))
Antikörper
Recommended dilution: Immunofluorescence: 1:10-1:50

Type: Primary
Antigen: Treponema pallidum
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR