Anti-TREML2 Rat monoclonal antibody PE (Phycoerythrin) [clone: 12K189]

USBIT8282-40DEA 489 EUR
USBIT8282-40D
Anti-TREML2 Rat monoclonal antibody PE (Phycoerythrin) [clone: 12K189]
Do not freeze!

Type: Primary
Antigen: TREML2 (triggering receptor expressed on myeloid cells-like 2)
Clonality: monoclonal
Clone: 12K189
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR