Anti-TREML2 Sheep Polyclonal Antibody

Lieferant: United States Biological
USBIT8282-40AEA 793 EUR
USBIT8282-40A
Anti-TREML2 Sheep Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 1 ?g/ml

Type: Primary
Antigen: TREML2 (triggering receptor expressed on myeloid cells-like 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR