Anti-TREML1 Rat Monoclonal Antibody [clone: 8H131]

Lieferant: United States Biological
USBIT8282-38EA 742 EUR
USBIT8282-38
Anti-TREML1 Rat Monoclonal Antibody [clone: 8H131]
Antikörper
Recommended dilution: Western Blot: 1-2?g/ml

Type: Primary
Antigen: TREML1 (triggering receptor expressed on myeloid cells-like 1)
Clonality: monoclonal
Clone: 8H131
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR