Anti-TREM2 Goat Polyclonal Antibody

Lieferant: United States Biological
USBIT8282-12EA 551 EUR
USBIT8282-12
Anti-TREM2 Goat Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:16000; Western Blot: 0. 3-1 ?g/ml

Type: Primary
Antigen: TREM2 (triggering receptor expressed on myeloid cells 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR