Anti-TREM1 Rat Monoclonal Antibody (FITC (Fluorescein)) [clone: 10A14]

Lieferant: United States Biological
USBIT8282-09EEA 533 EUR
USBIT8282-09E
Anti-TREM1 Rat Monoclonal Antibody (FITC (Fluorescein)) [clone: 10A14]
Antikörper
Recommended dilution: Flow Cytometry:1:10

Type: Primary
Antigen: TREM1 (triggering receptor expressed on myeloid cells 1)
Clonality: monoclonal
Clone: 10A14
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR