Anti-TREM1 Mouse Monoclonal Antibody [clone: 9F96]
Lieferant: United States Biological
USBIT8282-09EA 330 EUR
USBIT8282-09
Anti-TREM1 Mouse Monoclonal Antibody [clone: 9F96]
Antikörper
Recommended dilution: Western Blot: 1 ?g/ml
Type: Primary
Antigen: TREM1 (triggering receptor expressed on myeloid cells 1)
Clonality: monoclonal
Clone: 9F96
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human