Anti-TREK1 Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBIT8280-01AEA 539 EUR
USBIT8280-01A
Anti-TREK1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TREK1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR