Anti-TRBP2 Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBIT8269-01EEA 633 EUR
USBIT8269-01E
Anti-TRBP2 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Immunochemistry: 10 ?g/ml

Type: Primary
Antigen: TRBP2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR