Anti-TF Goat Polyclonal Antibody

Lieferant: United States Biological
USBIT8199-11AEA 327 EUR
USBIT8199-11A
Anti-TF Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TF (transferrin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR