Anti-ERLIN2 Goat Polyclonal Antibody

Lieferant: United States Biological
USBIS5402-01AEA 531 EUR
USBIS5402-01A
Anti-ERLIN2 Goat Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:2000; Western Blot: 0. 3-1 ?g/ml

Type: Primary
Antigen: ERLIN2 (ER lipid raft associated 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR