Anti-PSMA5 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Lieferant: United States Biological
USBIP9076-07C-PEEA 852 EUR
USBIP9076-07C-PE
Anti-PSMA5 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:50-1:100; Immunochemistry: 1:10-1:50
Do not freeze!
Type: Primary
Antigen: PSMA5 (proteasome (prosome, macropain) subunit, alpha type, 5)
Clonality: polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human