Anti-Procalcitonin Mouse Monoclonal Antibody [clone: 5E754]
Lieferant: United States Biological
USBIP9005-25DEA 472 EUR
USBIP9005-25D
Anti-Procalcitonin Mouse Monoclonal Antibody [clone: 5E754]
Antikörper
Type: Primary
Antigen: Procalcitonin
Clonality: monoclonal
Clone: 5E754
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human