Anti-PPARG Mouse Monoclonal Antibody [clone: 3A4A9, 1E6A1]
Lieferant: United States Biological
USBIP5505-06JEA 651 EUR
USBIP5505-06J
Anti-PPARG Mouse Monoclonal Antibody [clone: 3A4A9, 1E6A1]
Antikörper
Recommended dilution: ELISA: 1:10000; Western Blot: 1:500-1:2000; Immunochemistry: 1:200-1:400
Type: Primary
Antigen: PPARG (peroxisome proliferator-activated receptor gamma)
Clonality: monoclonal
Clone: 3A4A9, 1E6A1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human