Anti-PLCG1 Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBIP4074-43GEA 391 EUR
USBIP4074-43G
Anti-PLCG1 Rabbit Polyclonal Antibody
Antikörper
  • Recommended dilution: Western Blot: 1:500-1:1000

Type: Primary
Antigen: PLCG1 (phospholipase C, gamma 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR