Anti-PTCHD2 Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBIP3113-82T1EA 776 EUR
USBIP3113-82T1
Anti-PTCHD2 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: PTCHD2 (patched domain containing 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR