Anti-Nurim Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBIN8385-50AEA 480 EUR
USBIN8385-50A
Anti-Nurim Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Nurim
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR