Anti-BNIP1 Rabbit Polyclonal Antibody (PE (Phycoerythrin))

Lieferant: United States Biological
USBIN2567-80A-PEEA 852 EUR
USBIN2567-80A-PE
Anti-BNIP1 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-1:500; Immunochemistry: 1:50-1:100
Do not freeze!

Type: Primary
Antigen: BNIP1 (BCL2/adenovirus E1B 19kDa interacting protein 1)
Clonality: polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: BH3 domain
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR