Anti-Mucin 1 Mouse Monoclonal Antibody [clone: VU-4H5]

Lieferant: United States Biological
USBIM4700-11WEA 547 EUR
USBIM4700-11W
Anti-Mucin 1 Mouse Monoclonal Antibody [clone: VU-4H5]
Antikörper
Recommended dilution: Immunochemistry: 1:1000

Type: Primary
Antigen: Mucin 1
Clonality: monoclonal
Clone: VU-4H5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR