Anti-MAP4K1 Rabbit Polyclonal Antibody (PE (Phycoerythrin))

Lieferant: United States Biological
USBIM3949-71A-PEEA 871 EUR
USBIM3949-71A-PE
Anti-MAP4K1 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Antikörper
Recommended dilution: FLISA: 1:1000 Dot Blot: 1:500
Do not freeze!

Type: Primary
Antigen: MAP4K1 (mitogen-activated protein kinase kinase kinase kinase 1)
Clonality: polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR