Anti-MAPKAP1 Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBIM2354-88NEA 512 EUR
USBIM2354-88N
Anti-MAPKAP1 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 1-2 ?g/ml

Type: Primary
Antigen: MAPKAP1 (mitogen-activated protein kinase associated protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR