Anti-Ly49I Mouse Monoclonal Antibody (Biotin) [clone: 5K29]

Lieferant: United States Biological
USBIL7732-35BEA 463 EUR
USBIL7732-35B
Anti-Ly49I Mouse Monoclonal Antibody (Biotin) [clone: 5K29]
Antikörper
Type: Primary
Antigen: Ly49I
Clonality: monoclonal
Clone: 5K29
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR