Anti-Laminin A4 Goat Polyclonal Antibody (Biotin)

Lieferant: United States Biological
USBIL1225-49BEA 857 EUR
USBIL1225-49B
Anti-Laminin A4 Goat Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: Laminin A4
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR