Anti-LTF Goat Polyclonal Antibody (Biotin)
Lieferant: United States Biological
USBIL1021-82BEA 608 EUR
USBIL1021-82B
Anti-LTF Goat Polyclonal Antibody (Biotin)
Antikörper
Recommended dilution: ELISA: 1:2000-1:10000
Type: Primary
Antigen: LTF (lactotransferrin)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Monkey