Anti-KCNE2 Goat Polyclonal Antibody

Lieferant: United States Biological
USBIK0136-32EA 628 EUR
USBIK0136-32
Anti-KCNE2 Goat Polyclonal Antibody
Antikörper
Recommended dilution: Immunochemistry: 3. 75-7. 5 ?g/ml

Type: Primary
Antigen: KCNE2 (potassium voltage-gated channel, Isk-related family, member 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR