Anti-ELAVL1 Rabbit polyclonal antibody

Lieferant: United States Biological
USBIH7953-34EA 688 EUR
USBIH7953-34
Anti-ELAVL1 Rabbit polyclonal antibody
Antikörper
Type: Primary
Antigen: ELAVL1 (ELAV like RNA binding protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR