Anti-Histone H3, trimethyl Rabbit polyclonal antibody
Lieferant: United States Biological
USBIH5110-11H3EA 721 EUR
USBIH5110-11H3
Anti-Histone H3, trimethyl Rabbit polyclonal antibody
Antikörper
Recommended dilution: Western Blot: 1:1000-1:6000
Type: Primary
Antigen: Histone H3, trimethyl
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human