Anti-Hydroxybutyrate Dehydrogenase Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBIH0529-17AEA 654 EUR
USBIH0529-17A
Anti-Hydroxybutyrate Dehydrogenase Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Hydroxybutyrate Dehydrogenase
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR