Anti-GUCY1A3 Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBIG9806-24NEA 587 EUR
USBIG9806-24N
Anti-GUCY1A3 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500; Immunochemistry: 1:50-1:100

Type: Primary
Antigen: GUCY1A3 (guanylate cyclase 1, soluble, alpha 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR