Anti-Golgi Protein 58K Goat polyclonal antibody

Lieferant: United States Biological
USBIG8577-50GEA 770 EUR
USBIG8577-50G
Anti-Golgi Protein 58K Goat polyclonal antibody
Antikörper
Recommended dilution: ELISA: 1:64000; Western Blot: 0. 03-0.5 ?g/ml

Type: Primary
Antigen: Golgi Protein 58K
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR