Anti-Glycogen Phosphorylase, Muscle Rabbit polyclonal antibody

Lieferant: United States Biological
USBIG8170-16MEA 699 EUR
USBIG8170-16M
Anti-Glycogen Phosphorylase, Muscle Rabbit polyclonal antibody
Antikörper
Recommended dilution: Immunochemistry: 20 ?g/ml

Type: Primary
Antigen: Glycogen Phosphorylase, Muscle
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR