Anti-MAP4K2 Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBIG3009-02EA 587 EUR
USBIG3009-02
Anti-MAP4K2 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500; Immunochemistry: 1:50-1:100

Type: Primary
Antigen: MAP4K2 (mitogen-activated protein kinase kinase kinase kinase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR