Anti-FOXP3 Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBIF9050-10SEA 507 EUR
USBIF9050-10S
Anti-FOXP3 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 2-5 ?g/ml

Type: Primary
Antigen: FOXP3 (forkhead box P3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, Chicken, Horse, Human, Pig, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR