Anti-Fatty Acid Binding Protein, Heart Mouse monoclonal antibody [clone: 12H23]

Lieferant: United States Biological
USBIF0019-76MEA 883 EUR
USBIF0019-76M
Anti-Fatty Acid Binding Protein, Heart Mouse monoclonal antibody [clone: 12H23]
Antikörper
Type: Primary
Antigen: Fatty Acid Binding Protein, Heart
Clonality: monoclonal
Clone: 12H23
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR