Anti-PTK2 Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBIF0019-55JEA 575 EUR
USBIF0019-55J
Anti-PTK2 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 1:1000

Type: Primary
Antigen: PTK2 (protein tyrosine kinase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Chicken, Frog, Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR