Anti-EHMT1 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Lieferant: United States Biological
USBIE8947-50F-PEEA 852 EUR
USBIE8947-50F-PE
Anti-EHMT1 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:50-1:100
Do not freeze!
Type: Primary
Antigen: EHMT1 (euchromatic histone-lysine N-methyltransferase 1)
Clonality: polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human