Anti-ECE1 Rabbit Polyclonal Antibody (Biotin)

Lieferant: United States Biological
USBIE3650-01A-BIOTEA 791 EUR
USBIE3650-01A-BIOT
Anti-ECE1 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: ECE1 (endothelin converting enzyme 1)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR