Anti-EDIL3 Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBIE2204-30EA 584 EUR
USBIE2204-30
Anti-EDIL3 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500; Immunochemistry: 1:50-1:100; Flow Cytometry: 1:10-1:50

Type: Primary
Antigen: EDIL3 (EGF-like repeats and discoidin I-like domains 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR