Anti-Dopamine Rat Polyclonal Antibody (Unconjugated)

Lieferant: United States Biological
USBID9000-01QEA 760 EUR
USBID9000-01Q
Anti-Dopamine Rat Polyclonal Antibody (Unconjugated)
Antikörper
Type: Primary
Antigen: Dopamine
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR