Anti-Cytochrome P450 2B1 Mouse Monoclonal Antibody [clone: 9L780]
Lieferant: United States Biological
USBIC9095-15B5EA 506 EUR
USBIC9095-15B5
Anti-Cytochrome P450 2B1 Mouse Monoclonal Antibody [clone: 9L780]
Antikörper
Recommended dilution: Western Blot: 1:1000
Type: Primary
Antigen: Cytochrome P450 2B1
Clonality: Monoclonal
Clone: 9L780
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Rat