Anti-C1QTNF1 Mouse monoclonal antibody PE (Phycoerythrin) [clone: 10B2623]

USBIC7948-18D-PEEA 0 EUR
USBIC7948-18D-PE
Anti-C1QTNF1 Mouse monoclonal antibody PE (Phycoerythrin) [clone: 10B2623]
Type: Primary
Antigen: C1QTNF1 (C1q and tumor necrosis factor related protein 1)
Clonality: Monoclonal
Clone: 10B2623
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR