Anti-CTBS Mouse monoclonal antibody FITC (Fluorescein) [clone: 10B2310]

USBIC7947-28B-FITCEA 0 EUR
USBIC7947-28B-FITC
Anti-CTBS Mouse monoclonal antibody FITC (Fluorescein) [clone: 10B2310]
Type: Primary
Antigen: CTBS (chitobiase, di-N-acetyl-)
Clonality: Monoclonal
Clone: 10B2310
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR