Anti-CSTF3 Mouse monoclonal antibody HRP (Horseradish Peroxidase) [clone: 10B2620]

USBIC7946-70L-HRPEA 0 EUR
USBIC7946-70L-HRP
Anti-CSTF3 Mouse monoclonal antibody HRP (Horseradish Peroxidase) [clone: 10B2620]
Type: Primary
Antigen: CSTF3 (cleavage stimulation factor, 3' pre-RNA, subunit 3, 77kDa)
Clonality: Monoclonal
Clone: 10B2620
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR