Anti-Cleavage Stimulation Factor 2 Rabbit Polyclonal Antibody (Biotin)

Lieferant: United States Biological
USBIC7946-70C-BIOTEA 0 EUR
USBIC7946-70C-BIOT
Anti-Cleavage Stimulation Factor 2 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: Cleavage Stimulation Factor 2
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR