Anti-Cleavage Stimulation Factor 2 Rabbit polyclonal antibody

Lieferant: United States Biological
USBIC7946-70CEA 0 EUR
USBIC7946-70C
Anti-Cleavage Stimulation Factor 2 Rabbit polyclonal antibody
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500

Type: Primary
Antigen: Cleavage Stimulation Factor 2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR