Anti-CSN3 Mouse monoclonal antibody AP (Alkaline Phosphatase) [clone: 10B2309]

USBIC7945-70E-APEA 0 EUR
USBIC7945-70E-AP
Anti-CSN3 Mouse monoclonal antibody AP (Alkaline Phosphatase) [clone: 10B2309]
Type: Primary
Antigen: CSN3 (casein kappa)
Clonality: Monoclonal
Clone: 10B2309
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR